Product Details

SNP ID
rs200258703
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100093122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGGTGGCAAATATCACATCTGCT[A/G]GTCTCTGAGTCCTGAAGGAAATGAG
Phenotype
MIM: 602296 MIM: 614729 MIM: 600592 MIM: 612983 MIM: 612150 MIM: 612984
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
AP4M1 PubMed Links
Additional Information
For this assay, SNP(s) [rs1130958] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
AP4M1
Gene Name
adaptor related protein complex 4 mu 1 subunit
There are no transcripts associated with this gene.

Gene
COPS6
Gene Name
COP9 signalosome subunit 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006833.4 2616 Intron NP_006824.2
Gene
MCM7
Gene Name
minichromosome maintenance complex component 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278595.1 2616 Missense Mutation CCA,CTA P481L NP_001265524.1
NM_005916.4 2616 Missense Mutation CCA,CTA P657L NP_005907.3
NM_182776.2 2616 Missense Mutation CCA,CTA P481L NP_877577.1
XM_005250348.3 2616 Missense Mutation CCA,CTA P550L XP_005250405.1
XM_017012217.1 2616 Missense Mutation CCA,CTA P550L XP_016867706.1
Gene
MIR106B
Gene Name
microRNA 106b
There are no transcripts associated with this gene.

Gene
MIR25
Gene Name
microRNA 25
There are no transcripts associated with this gene.

Gene
MIR93
Gene Name
microRNA 93
There are no transcripts associated with this gene.

View Full Product Details