Product Details

SNP ID
rs200186151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:94910941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGGATGCTCACAAGAGTAATGC[A/G]ACTCCAGTACCAGAAGTGGCTTCTA
Phenotype
MIM: 602468
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP1R9A PubMed Links

Gene Details

Gene
PPP1R9A
Gene Name
protein phosphatase 1 regulatory subunit 9A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166160.1 1110 Silent Mutation GCA,GCG A276A NP_001159632.1
NM_001166161.1 1110 Silent Mutation GCA,GCG A276A NP_001159633.1
NM_001166162.1 1110 Silent Mutation GCA,GCG A276A NP_001159634.1
NM_001166163.1 1110 Silent Mutation GCA,GCG A276A NP_001159635.1
NM_017650.2 1110 Silent Mutation GCA,GCG A276A NP_060120.2
XM_011516380.1 1110 Silent Mutation GCA,GCG A276A XP_011514682.1
XM_011516381.1 1110 Silent Mutation GCA,GCG A276A XP_011514683.1
XM_011516382.2 1110 Silent Mutation GCA,GCG A276A XP_011514684.1
XM_011516383.1 1110 Silent Mutation GCA,GCG A276A XP_011514685.1
XM_011516389.1 1110 Silent Mutation GCA,GCG A276A XP_011514691.1
XM_017012394.1 1110 Silent Mutation GCA,GCG A276A XP_016867883.1
XM_017012395.1 1110 Silent Mutation GCA,GCG A276A XP_016867884.1
XM_017012396.1 1110 Silent Mutation GCA,GCG A276A XP_016867885.1
XM_017012397.1 1110 Silent Mutation GCA,GCG A276A XP_016867886.1
XM_017012398.1 1110 Silent Mutation GCA,GCG A276A XP_016867887.1
XM_017012399.1 1110 Silent Mutation GCA,GCG A276A XP_016867888.1
XM_017012400.1 1110 Silent Mutation GCA,GCG A276A XP_016867889.1
XM_017012401.1 1110 Silent Mutation GCA,GCG A276A XP_016867890.1
XM_017012402.1 1110 Silent Mutation GCA,GCG A276A XP_016867891.1
XM_017012403.1 1110 Silent Mutation GCA,GCG A276A XP_016867892.1
XM_017012404.1 1110 Silent Mutation GCA,GCG A276A XP_016867893.1
XM_017012405.1 1110 Silent Mutation GCA,GCG A276A XP_016867894.1
XM_017012406.1 1110 Silent Mutation GCA,GCG A276A XP_016867895.1
XM_017012407.1 1110 Silent Mutation GCA,GCG A276A XP_016867896.1
XM_017012408.1 1110 Intron XP_016867897.1
XM_017012409.1 1110 Silent Mutation GCA,GCG A276A XP_016867898.1
XM_017012410.1 1110 UTR 5 XP_016867899.1
XM_017012411.1 1110 Intron XP_016867900.1

View Full Product Details