Product Details

SNP ID
rs201728682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:75073695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCCCAGCCCCGGCCCGCTCAGCC[A/G]CCGCCCCAGCCGAGCCCCAGCCACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RCC1L PubMed Links

Gene Details

Gene
RCC1L
Gene Name
RCC1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281441.1 187 Missense Mutation CGG,TGG R15W NP_001268370.1
NM_030798.4 187 Missense Mutation CGG,TGG R15W NP_110425.2
NM_148842.2 187 Missense Mutation CGG,TGG R15W NP_683682.1

View Full Product Details