Product Details

SNP ID
rs201716266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:2235329 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGCAGGATGTCTGGGGTCACGCT[A/G]AGAAGGGTCAGGCACAGGCTGATGA
Phenotype
MIM: 602686 MIM: 606906 MIM: 600312
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAD1L1 PubMed Links

Gene Details

Gene
MAD1L1
Gene Name
MAD1 mitotic arrest deficient like 1
There are no transcripts associated with this gene.

Gene
MRM2
Gene Name
mitochondrial rRNA methyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013393.1 563 Silent Mutation CTC,CTT L178L NP_037525.1
Gene
NUDT1
Gene Name
nudix hydrolase 1
There are no transcripts associated with this gene.

View Full Product Details