Product Details
- SNP ID
-
rs201813130
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:149104102 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGAGCTGAGACTGATGCGTGTAG[A/G]TCTTGTCACACTCACCGCACGCGAA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF425
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11525060] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF425
- Gene Name
- zinc finger protein 425
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001001661.2 |
1902 |
Missense Mutation |
ACC,ATC |
T590I |
NP_001001661.1 |
View Full Product Details