Product Details

SNP ID
rs201212455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73308761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGCGGCTCTCCACGCCGGCCAA[C/T]ACGCCTGCAGCTGCAGCATACAGCC
Phenotype
MIM: 615732 MIM: 612548
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NSUN5 PubMed Links

Gene Details

Gene
NSUN5
Gene Name
NOP2/Sun RNA methyltransferase family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168347.2 Intron NP_001161819.1
NM_001168348.2 Intron NP_001161820.1
NM_018044.4 Intron NP_060514.1
NM_148956.3 Intron NP_683759.1
XM_005277604.4 Intron XP_005277661.1
Gene
TRIM50
Gene Name
tripartite motif containing 50
There are no transcripts associated with this gene.

View Full Product Details