Product Details
- SNP ID
-
rs201326665
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:100723038 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTCCACTCCGAACAATCACTGCT[C/G]ACACTTTCCGCAAACTCTTCCGAGT
- Phenotype
-
MIM: 133170
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
EPO
PubMed Links
Gene Details
- Gene
- EPO
- Gene Name
- erythropoietin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000799.2 |
668 |
Missense Mutation |
CAC,GAC |
H163D |
NP_000790.2 |
View Full Product Details