Product Details
- SNP ID
-
rs201196772
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:135571137 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTATGGGGTCCTTACAAAGACATT[G/T]GGCATAAAGTGGGAAATGCTCTTTG
- Phenotype
-
MIM: 614352
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NUP205
PubMed Links
Gene Details
- Gene
- NUP205
- Gene Name
- nucleoporin 205
View Full Product Details