Product Details

SNP ID
rs201983608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:23680267 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCGGCGCGGCGGCGGTGGACGAG[G/T]ACCTGGAGTACCGGAGGTGAGGCTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM221A PubMed Links

Gene Details

Gene
FAM221A
Gene Name
family with sequence similarity 221 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127364.2 149 Missense Mutation GAC,TAC D17Y NP_001120836.1
NM_001127365.2 149 Missense Mutation GAC,TAC D17Y NP_001120837.1
NM_001300932.1 149 Missense Mutation GAC,TAC D17Y NP_001287861.1
NM_199136.4 149 Missense Mutation GAC,TAC D17Y NP_954587.2
XM_011515369.2 149 Missense Mutation GAC,TAC D17Y XP_011513671.1
XM_011515370.2 149 Missense Mutation GAC,TAC D17Y XP_011513672.1
XM_011515371.2 149 Missense Mutation GAC,TAC D17Y XP_011513673.1
XM_011515372.2 149 Intron XP_011513674.1
XM_017012134.1 149 Missense Mutation GAC,TAC D17Y XP_016867623.1
XM_017012135.1 149 Intron XP_016867624.1

View Full Product Details