Product Details

SNP ID
rs201633210
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100738528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTGACTGGACCTGGATCCCAAC[A/T]GCCTCCGGGGCCAAGTGGACTCAGA
Phenotype
MIM: 602372
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZAN PubMed Links

Gene Details

Gene
ZAN
Gene Name
zonadhesin (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003386.2 846 Silent Mutation ACA,ACT T227T NP_003377.2
NM_173059.2 846 Silent Mutation ACA,ACT T227T NP_775082.2
XM_011516555.2 846 Silent Mutation ACA,ACT T227T XP_011514857.1
XM_017012581.1 846 Intron XP_016868070.1

View Full Product Details