Product Details

SNP ID
rs199552953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:102446703 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAACTCCATCAGCGAGTCCCCGC[A/G]TGAGCGCATGCACCCCTACATCGAG
Phenotype
MIM: 613302 MIM: 610929
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALKBH4 PubMed Links

Gene Details

Gene
ALKBH4
Gene Name
alkB homolog 4, lysine demethylase
There are no transcripts associated with this gene.

Gene
ORAI2
Gene Name
ORAI calcium release-activated calcium modulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126340.2 561 Missense Mutation CAT,CGT H139R NP_001119812.1
NM_001271818.1 561 Missense Mutation CAT,CGT H139R NP_001258747.1
NM_001271819.1 561 Missense Mutation CAT,CGT H62R NP_001258748.1
NM_032831.3 561 Missense Mutation CAT,CGT H139R NP_116220.1

View Full Product Details