Product Details

SNP ID
rs202201309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:37937327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCCATGCATAGCTTCTTGGGGT[C/T]CCAGACAGAGTATTCATAAACTGTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GOT1L1 PubMed Links

Gene Details

Gene
GOT1L1
Gene Name
glutamic-oxaloacetic transaminase 1-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152413.2 942 Missense Mutation AAC,GAC N157D NP_689626.2
XM_005273399.3 942 Missense Mutation AAC,GAC N157D XP_005273456.1
XM_006716285.3 942 Missense Mutation AAC,GAC N157D XP_006716348.1
XM_011544391.2 942 Intron XP_011542693.1

View Full Product Details