Product Details
- SNP ID
-
rs200872639
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:89758090 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACGGGGAGGCCATCTGCAGCGCCCT[G/T]CCCACCATTCCCTACCACAAACTCG
- Phenotype
-
MIM: 603455
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101929709
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2293809] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101929709
- Gene Name
- uncharacterized LOC101929709
There are no transcripts associated with this gene.
- Gene
- RIPK2
- Gene Name
- receptor interacting serine/threonine kinase 2
View Full Product Details