Product Details
- SNP ID
-
rs201108325
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:23844836 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTCCTCCTCACCTCGGAGGTTCCT[C/G]AGGAGGACTTTCAGCTTCTGCGGCT
- Phenotype
-
MIM: 601185
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
STC1
PubMed Links
Gene Details
- Gene
- STC1
- Gene Name
- stanniocalcin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003155.2 |
962 |
Silent Mutation |
CTC,CTG |
L226L |
NP_003146.1 |
View Full Product Details