Product Details

SNP ID
rs199761990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:48919975 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAGATCTTGCAAACACAAGGTAA[C/T]GTGTGGGTCCGAATATGCATCTTCA
Phenotype
MIM: 602150
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNAI2 PubMed Links

Gene Details

Gene
SNAI2
Gene Name
snail family transcriptional repressor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003068.4 721 Silent Mutation ACA,ACG T182T NP_003059.1

View Full Product Details