Product Details

SNP ID
rs200420761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:109087519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACTTTAAGCTTCTAAAGATGGAA[A/G]ACGAGACAGTCAGTGAACTGAACCA
Phenotype
MIM: 188545
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRHR PubMed Links

Gene Details

Gene
TRHR
Gene Name
thyrotropin releasing hormone receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003301.5 96 Missense Mutation AAC,GAC N3D NP_003292.1
XM_011517263.2 96 Missense Mutation AAC,GAC N3D XP_011515565.1

View Full Product Details