Product Details
- SNP ID
-
rs200726525
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:8703178 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCGGCCCAAGGCCTACACCAACTC[A/G]GTGGACGTCCTCGACGGGGAGGGGT
- Phenotype
-
MIM: 609203
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLDN23
PubMed Links
Gene Details
- Gene
- CLDN23
- Gene Name
- claudin 23
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_194284.2 |
1023 |
Silent Mutation |
TCA,TCG |
S260S |
NP_919260.2 |
View Full Product Details