Product Details

SNP ID
rs200115790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:85442101 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGAGGGGGTCCTCTCCCTGGACC[C/T]TACCGACTTCGCCAGTTTCATCTTC
Phenotype
MIM: 114750
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CA3 PubMed Links

Gene Details

Gene
CA3
Gene Name
carbonic anhydrase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005181.3 344 Silent Mutation CCC,CCT P87P NP_005172.1
Gene
CA3-AS1
Gene Name
CA3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details