Product Details
- SNP ID
-
rs200692511
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:112997576 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGTTTTTCTCCAGTATGAATTCT[G/T]TGATGTTGAATGAAGGCTGGGGTAT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF883
PubMed Links
Gene Details
- Gene
- ZNF883
- Gene Name
- zinc finger protein 883
View Full Product Details