Product Details
- SNP ID
-
rs201484497
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:93447660 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGTGTTGGTGGTTTTGTGGTTTTC[C/T]TTGAGACTTTGTTATTGACAGAACA
- Phenotype
-
MIM: 612265
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM120A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10821128] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM120A
- Gene Name
- family with sequence similarity 120A
There are no transcripts associated with this gene.
- Gene
- FAM120AOS
- Gene Name
- family with sequence similarity 120A opposite strand
View Full Product Details