Product Details

SNP ID
rs200204592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34254427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCTGCGGTGGGCCCGTGGCAGG[G/T]GAGATAGAGCTGCAGCTGGCTCCTC
Phenotype
MIM: 613747 MIM: 609787
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIF24 PubMed Links

Gene Details

Gene
KIF24
Gene Name
kinesin family member 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194313.2 2784 Missense Mutation NP_919289.2
XM_005251451.2 2784 Missense Mutation XP_005251508.1
XM_006716764.2 2784 Missense Mutation XP_006716827.1
XM_011517861.1 2784 Missense Mutation XP_011516163.1
XM_011517863.2 2784 Missense Mutation XP_011516165.1
XM_017014697.1 2784 Missense Mutation XP_016870186.1
XM_017014698.1 2784 Missense Mutation XP_016870187.1
Gene
UBAP1
Gene Name
ubiquitin associated protein 1
There are no transcripts associated with this gene.

View Full Product Details