Product Details

SNP ID
rs200496124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:121567198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGGCCCACACGCCATGGAGCCC[A/G]ACTCCCTTCTGGACCAAGACGACTC
Phenotype
MIM: 609205
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DAB2IP PubMed Links

Gene Details

Gene
DAB2IP
Gene Name
DAB2 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032552.3 316 Missense Mutation AAC,GAC N4D NP_115941.2
NM_138709.2 316 Intron NP_619723.1
XM_005251719.4 316 Intron XP_005251776.1
XM_005251721.1 316 Missense Mutation AAC,GAC N4D XP_005251778.1
XM_005251724.4 316 Intron XP_005251781.1
XM_011518264.2 316 Intron XP_011516566.1
XM_011518265.2 316 Intron XP_011516567.1
XM_011518266.2 316 Intron XP_011516568.1
XM_011518267.2 316 Intron XP_011516569.1
XM_011518270.2 316 Intron XP_011516572.1
XM_011518271.2 316 Intron XP_011516573.1
XM_017014298.1 316 Intron XP_016869787.1
XM_017014299.1 316 Intron XP_016869788.1
XM_017014300.1 316 Intron XP_016869789.1

View Full Product Details