Product Details
- SNP ID
-
rs200577968
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:3225153 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGGAGGCTTGGTTGCACGCCAGT[C/T]TCGAGAACAGGCTGCATGCAGCCCA
- Phenotype
-
MIM: 601337
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RFX3
PubMed Links
Gene Details
- Gene
- RFX3
- Gene Name
- regulatory factor X3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001282116.1 |
2460 |
Silent Mutation |
GAA,GAG |
E713E |
NP_001269045.1 |
NM_001282117.1 |
2460 |
Intron |
|
|
NP_001269046.1 |
NM_002919.3 |
2460 |
Intron |
|
|
NP_002910.1 |
NM_134428.2 |
2460 |
Silent Mutation |
GAA,GAG |
E713E |
NP_602304.1 |
XM_006716840.1 |
2460 |
Silent Mutation |
GAA,GAG |
E744E |
XP_006716903.1 |
XM_006716843.1 |
2460 |
Silent Mutation |
GAA,GAG |
E734E |
XP_006716906.1 |
XM_006716844.1 |
2460 |
Silent Mutation |
GAA,GAG |
E734E |
XP_006716907.1 |
XM_006716845.2 |
2460 |
Silent Mutation |
GAA,GAG |
E734E |
XP_006716908.1 |
XM_006716847.3 |
2460 |
Silent Mutation |
GAA,GAG |
E456E |
XP_006716910.1 |
XM_011517998.1 |
2460 |
Silent Mutation |
GAA,GAG |
E734E |
XP_011516300.1 |
XM_011517999.1 |
2460 |
Silent Mutation |
GAA,GAG |
E734E |
XP_011516301.1 |
XM_011518000.1 |
2460 |
Silent Mutation |
GAA,GAG |
E709E |
XP_011516302.1 |
XM_011518001.1 |
2460 |
Silent Mutation |
GAA,GAG |
E651E |
XP_011516303.1 |
XM_017015000.1 |
2460 |
Silent Mutation |
GAA,GAG |
E734E |
XP_016870489.1 |
XM_017015001.1 |
2460 |
Silent Mutation |
GAA,GAG |
E688E |
XP_016870490.1 |
XM_017015002.1 |
2460 |
Silent Mutation |
GAA,GAG |
E465E |
XP_016870491.1 |
View Full Product Details