Product Details

SNP ID
rs200413372
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128721301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCAGGCCTCGGTCGAAGGGGTT[A/G]CTGGGGCGCTGGCGGAGATAGGCGA
Phenotype
MIM: 610714
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100506100 PubMed Links

Gene Details

Gene
LOC100506100
Gene Name
uncharacterized LOC100506100
There are no transcripts associated with this gene.

Gene
PKN3
Gene Name
protein kinase N3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317926.1 872 Intron NP_001304855.1
NM_013355.4 872 Intron NP_037487.2
XM_005251946.3 872 Intron XP_005252003.1
XM_006717080.2 872 Intron XP_006717143.1
XM_017014649.1 872 Intron XP_016870138.1
XM_017014650.1 872 Intron XP_016870139.1
Gene
ZDHHC12
Gene Name
zinc finger DHHC-type containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318015.1 872 Silent Mutation AGC,AGT S283S NP_001304944.1
NM_001318016.1 872 Nonsense Mutation CAA,TAA Q238* NP_001304945.1
NM_001318020.1 872 Missense Mutation GCA,GTA A221V NP_001304949.1
NM_001318023.1 872 Silent Mutation AGC,AGT S227S NP_001304952.1
NM_032799.4 872 Silent Mutation AGC,AGT S228S NP_116188.2
XM_011519116.2 872 Missense Mutation GCA,GTA A276V XP_011517418.1
Gene
ZER1
Gene Name
zyg-11 related cell cycle regulator
There are no transcripts associated with this gene.

View Full Product Details