Product Details

SNP ID
rs200906831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:99915317 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAATTCATTAAGATAGTAATCCC[A/C]ACAGACCTGGAAAGGTTAAGAAAGC
Phenotype
MIM: 604204
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
STX17 PubMed Links

Gene Details

Gene
STX17
Gene Name
syntaxin 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017919.2 545 Silent Mutation CCA,CCC P26P NP_060389.2
XM_011518820.2 545 Silent Mutation CCA,CCC P26P XP_011517122.1
XM_011518821.2 545 Silent Mutation CCA,CCC P26P XP_011517123.1
XM_011518823.2 545 Intron XP_011517125.1
XM_017014875.1 545 UTR 5 XP_016870364.1
Gene
STX17-AS1
Gene Name
STX17 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details