Product Details

SNP ID
rs200623737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137279875 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCCCGCGCGGAGAACCTGGCC[A/G]CGCAGCTCAGCTTCTACCGCGTGGC
Phenotype
MIM: 611180
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NELFB PubMed Links

Gene Details

Gene
NELFB
Gene Name
negative elongation factor complex member B
There are no transcripts associated with this gene.

Gene
TOR4A
Gene Name
torsin family 4 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017723.2 1303 Missense Mutation ACG,GCG T396A NP_060193.2

View Full Product Details