Product Details
- SNP ID
-
rs200104730
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:92846279 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCTTCTGAGTCCCCACACCATAA[C/T]TGTGGCATGGAGCACTCTACTTCCC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD19P
PubMed Links
Gene Details
- Gene
- ANKRD19P
- Gene Name
- ankyrin repeat domain 19, pseudogene
There are no transcripts associated with this gene.
- Gene
- ZNF484
- Gene Name
- zinc finger protein 484
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001007101.2 |
2642 |
Silent Mutation |
CAA,CAG |
Q800Q |
NP_001007102.1 |
NM_001261458.1 |
2642 |
Silent Mutation |
CAA,CAG |
Q838Q |
NP_001248387.1 |
NM_001261459.1 |
2642 |
Silent Mutation |
CAA,CAG |
Q800Q |
NP_001248388.1 |
NM_001261460.1 |
2642 |
Silent Mutation |
CAA,CAG |
Q800Q |
NP_001248389.1 |
NM_031486.2 |
2642 |
Silent Mutation |
CAA,CAG |
Q836Q |
NP_113674.1 |
XM_006717299.3 |
2642 |
Silent Mutation |
CAA,CAG |
Q815Q |
XP_006717362.1 |
XM_011519073.2 |
2642 |
Silent Mutation |
CAA,CAG |
Q868Q |
XP_011517375.1 |
XM_011519074.2 |
2642 |
Silent Mutation |
CAA,CAG |
Q853Q |
XP_011517376.1 |
XM_011519075.2 |
2642 |
Silent Mutation |
CAA,CAG |
Q851Q |
XP_011517377.1 |
XM_017015178.1 |
2642 |
Silent Mutation |
CAA,CAG |
Q815Q |
XP_016870667.1 |
XM_017015179.1 |
2642 |
Silent Mutation |
CAA,CAG |
Q815Q |
XP_016870668.1 |
XM_017015180.1 |
2642 |
Silent Mutation |
CAA,CAG |
Q815Q |
XP_016870669.1 |
XM_017015181.1 |
2642 |
Silent Mutation |
CAA,CAG |
Q800Q |
XP_016870670.1 |
View Full Product Details