Product Details

SNP ID
rs199765604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135665289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCGTCTCGGAGACTGACTACAGG[C/T]TGTTCATCACCTTCCACCTCCAGAA
Phenotype
MIM: 612903
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LCN9 PubMed Links

Gene Details

Gene
LCN9
Gene Name
lipocalin 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001676.1 352 Silent Mutation CTG,TTG L118L NP_001001676.1
XM_017014712.1 352 Missense Mutation CTG,TTG L118L XP_016870201.1

View Full Product Details