Product Details

SNP ID
rs199567728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128952806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGCAGAACTGAATAAACATTGG[C/T]GGCGATTGTTAGAGGGGCTTTCTTA
Phenotype
MIM: 610746 MIM: 615587
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DOLK PubMed Links

Gene Details

Gene
DOLK
Gene Name
dolichol kinase
There are no transcripts associated with this gene.

Gene
NUP188
Gene Name
nucleoporin 188
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015354.2 148 Missense Mutation CGG,TGG R41W NP_056169.1

View Full Product Details