Product Details

SNP ID
rs199881525
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:21481620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTCTTTGCTGGAAGATAATCAG[C/T]TTCAAATCTAGAGAGAAGATAGTGG
Phenotype
MIM: 615223 MIM: 616356
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFNE PubMed Links

Gene Details

Gene
IFNE
Gene Name
interferon epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176891.4 694 Silent Mutation AAA,AAG K25K NP_795372.1
Gene
MIR31HG
Gene Name
MIR31 host gene
There are no transcripts associated with this gene.

View Full Product Details