Product Details

SNP ID
rs199842194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137552443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCTGGAAAAGTACGGCTTCGAGC[C/T]CACACAGGAGGGAAAGCTCTTCCAG
Phenotype
MIM: 613210 MIM: 611846 MIM: 612122
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPH7 PubMed Links

Gene Details

Gene
DPH7
Gene Name
diphthamide biosynthesis 7
There are no transcripts associated with this gene.

Gene
MRPL41
Gene Name
mitochondrial ribosomal protein L41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032477.2 492 Missense Mutation CCC,CTC P121L NP_115866.1
Gene
PNPLA7
Gene Name
patatin like phospholipase domain containing 7
There are no transcripts associated with this gene.

View Full Product Details