Product Details

SNP ID
rs200075007
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:129322096 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCACACAGGGCTGTGAGCTCCAT[A/G]AGGGCAGGAACCACATGGCTGTTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf106 PubMed Links

Gene Details

Gene
C9orf106
Gene Name
chromosome 9 open reading frame 106
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012715.3 336 Missense Mutation AAG,GAG K95E NP_001012733.2

View Full Product Details