Product Details
- SNP ID
-
rs202212101
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:34402392 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCCAAGCCCCCATACACACCTGTC[C/T]GTGTCAAGGGCTACCAGTGGCTTCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C9orf24
PubMed Links
Gene Details
- Gene
- C9orf24
- Gene Name
- chromosome 9 open reading frame 24
There are no transcripts associated with this gene.
- Gene
- FAM219A
- Gene Name
- family with sequence similarity 219 member A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001184940.1 |
451 |
Silent Mutation |
ACA,ACG |
T113T |
NP_001171869.1 |
NM_001184941.1 |
451 |
Silent Mutation |
ACA,ACG |
T113T |
NP_001171870.1 |
NM_001184942.1 |
451 |
Silent Mutation |
ACA,ACG |
T102T |
NP_001171871.1 |
NM_001184943.1 |
451 |
Silent Mutation |
ACA,ACG |
T96T |
NP_001171872.1 |
NM_001184945.1 |
451 |
Silent Mutation |
ACA,ACG |
T85T |
NP_001171874.1 |
NM_147202.1 |
451 |
Silent Mutation |
ACA,ACG |
T96T |
NP_671735.1 |
XM_005251402.4 |
451 |
Silent Mutation |
ACA,ACG |
T72T |
XP_005251459.1 |
XM_011517797.2 |
451 |
Silent Mutation |
ACA,ACG |
T102T |
XP_011516099.1 |
XM_011517798.2 |
451 |
Silent Mutation |
ACA,ACG |
T85T |
XP_011516100.1 |
XM_011517799.2 |
451 |
Silent Mutation |
ACA,ACG |
T44T |
XP_011516101.1 |
View Full Product Details