Product Details

SNP ID
rs201800321
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:87728808 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAATCCTATCCATATGAGGCAA[C/T]AGTAAGTGGAGCTCCTTGTCATCAT
Phenotype
MIM: 116880
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTSL PubMed Links

Gene Details

Gene
CTSL
Gene Name
cathepsin L
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257971.1 1510 Missense Mutation ACA,ATA T207I NP_001244900.1
NM_001257972.1 1510 Missense Mutation ACA,ATA T207I NP_001244901.1
NM_001257973.1 1510 Missense Mutation ACA,ATA T25I NP_001244902.1
NM_001912.4 1510 Missense Mutation ACA,ATA T207I NP_001903.1
NM_145918.2 1510 Missense Mutation ACA,ATA T207I NP_666023.1
XM_005251716.3 1510 Missense Mutation ACA,ATA T207I XP_005251773.1
XM_011518263.1 1510 Missense Mutation ACA,ATA T207I XP_011516565.1
XM_017014293.1 1510 Missense Mutation ACA,ATA T207I XP_016869782.1

View Full Product Details