Product Details
- SNP ID
-
rs201905429
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:137278864 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCCGACGTCGGGACCGGGGCGCCC[A/G]GGCCGGGCTGCAGCCCCCGGGCACC
- Phenotype
-
MIM: 611180
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NELFB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150975361] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NELFB
- Gene Name
- negative elongation factor complex member B
There are no transcripts associated with this gene.
- Gene
- TOR4A
- Gene Name
- torsin family 4 member A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017723.2 |
292 |
Missense Mutation |
AGG,GGG |
R59G |
NP_060193.2 |
View Full Product Details