Product Details

SNP ID
rs201494183
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136984744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGAGTGACATCCGAGTGGCCTT[C/T]TCCGACTACCAGCACTTTGCCTTGC
Phenotype
MIM: 616315 MIM: 606533 MIM: 176803
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C9orf142 PubMed Links

Gene Details

Gene
C9orf142
Gene Name
chromosome 9 open reading frame 142
There are no transcripts associated with this gene.

Gene
CLIC3
Gene Name
chloride intracellular channel 3
There are no transcripts associated with this gene.

Gene
LCNL1
Gene Name
lipocalin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207510.3 822 Silent Mutation TTC,TTT F76F NP_997393.3
Gene
PTGDS
Gene Name
prostaglandin D2 synthase
There are no transcripts associated with this gene.

View Full Product Details