Product Details

SNP ID
rs201682816
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137008818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCTTGCTGAACACCTGGGCCAG[C/T]GAGATGTGCTCCGACTTGAGCTGGT
Phenotype
MIM: 600047
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCA2 PubMed Links

Gene Details

Gene
ABCA2
Gene Name
ATP binding cassette subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001606.4 7063 Silent Mutation NP_001597.2
NM_212533.2 7063 Silent Mutation NP_997698.1
XM_006716996.3 7063 Silent Mutation XP_006717059.1

View Full Product Details