Product Details

SNP ID
rs201725287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:124881884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTCAGTGTTATTCGTGACGGAA[A/G]GCGCCATGTTTGCCATCTCAGGTCC
Phenotype
MIM: 602502
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARPC5L PubMed Links

Gene Details

Gene
ARPC5L
Gene Name
actin related protein 2/3 complex subunit 5 like
There are no transcripts associated with this gene.

Gene
GOLGA1
Gene Name
golgin A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002077.3 2345 Missense Mutation CCT,CTT P679L NP_002068.1
XM_005251929.3 2345 Missense Mutation CCT,CTT P679L XP_005251986.1
XM_006717062.3 2345 Missense Mutation CCT,CTT P679L XP_006717125.1
XM_006717063.3 2345 Missense Mutation CCT,CTT P679L XP_006717126.1

View Full Product Details