Product Details

SNP ID
rs201565774
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136405163 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCGTGGTAGTCGCTTTCCTCCTT[C/G]ATCATTTTTGCTTCTAACTTCCGCT
Phenotype
MIM: 613036 MIM: 602777
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PMPCA PubMed Links

Gene Details

Gene
PMPCA
Gene Name
peptidase, mitochondrial processing alpha subunit
There are no transcripts associated with this gene.

Gene
SDCCAG3
Gene Name
serologically defined colon cancer antigen 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039707.1 922 Intron NP_001034796.1
NM_001039708.1 922 Intron NP_001034797.1
NM_006643.3 922 Intron NP_006634.3
XM_005266050.3 922 Intron XP_005266107.1
XM_005266051.3 922 Intron XP_005266108.1
XM_011518157.2 922 Missense Mutation ATC,ATG I278M XP_011516459.1
XM_017014218.1 922 Intron XP_016869707.1
XM_017014219.1 922 Intron XP_016869708.1
Gene
SNAPC4
Gene Name
small nuclear RNA activating complex polypeptide 4
There are no transcripts associated with this gene.

View Full Product Details