Product Details

SNP ID
rs201963967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:12775881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGGTGGCGGCGGCGGCGGCGGC[G/T]GCAGTAGCAGCAGCAGCTACTGCAG
Phenotype
MIM: 616130
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LURAP1L PubMed Links

Gene Details

Gene
LURAP1L
Gene Name
leucine rich adaptor protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203403.1 869 Missense Mutation GGC,TGC G56C NP_981948.1
XM_005251443.2 869 Missense Mutation GGC,TGC G56C XP_005251500.1
Gene
LURAP1L-AS1
Gene Name
LURAP1L antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details