Product Details

SNP ID
rs201672788
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34621540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTGGGAAGCTGGCACAGGCTGG[C/T]GGCGGGCAAAGAGGACACCTGGCAA
Phenotype
MIM: 607387
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ARID3C PubMed Links

Gene Details

Gene
ARID3C
Gene Name
AT-rich interaction domain 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017363.1 4243 Missense Mutation CAC,CGC H386R NP_001017363.1
XM_017014283.1 4243 Missense Mutation CAC,CGC H536R XP_016869772.1
Gene
DCTN3
Gene Name
dynactin subunit 3
There are no transcripts associated with this gene.

Gene
RPP25L
Gene Name
ribonuclease P/MRP subunit p25 like
There are no transcripts associated with this gene.

View Full Product Details