Product Details

SNP ID
rs199733827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:51743672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCGACTCGGCGCCCGATTGCTGG[A/G]ACCAGGTGGACATGGAATCCCCGGG
Phenotype
MIM: 300418
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSPT2 PubMed Links

Gene Details

Gene
GSPT2
Gene Name
G1 to S phase transition 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018094.4 288 Missense Mutation AAC,GAC N16D NP_060564.2

View Full Product Details