Product Details

SNP ID
rs199580855
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:118346541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGACACCGAGGAATTCTATGATG[C/T]CCCTGAAGATGTGCACCTAGGGGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDR44 PubMed Links

Gene Details

Gene
WDR44
Gene Name
WD repeat domain 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184965.1 469 Missense Mutation GCC,GTC A13V NP_001171894.1
NM_001184966.1 469 Missense Mutation GCC,GTC A13V NP_001171895.1
NM_019045.4 469 Missense Mutation GCC,GTC A13V NP_061918.3
XM_011531353.2 469 Missense Mutation GCC,GTC A13V XP_011529655.1
XM_017029600.1 469 UTR 5 XP_016885089.1

View Full Product Details