Product Details

SNP ID
rs199792420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:54751314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGACTGTGATGATCTGGAAGTAG[A/G]TGCGAGTCTGGTCCTTGTGGCCCGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITIH6 PubMed Links

Gene Details

Gene
ITIH6
Gene Name
inter-alpha-trypsin inhibitor heavy chain family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198510.2 3449 Missense Mutation ACC,ATC T1140I NP_940912.1

View Full Product Details