Product Details

SNP ID
rs200241333
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101276083 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATCTTATGGAGCTTTTTCTCCT[C/T]TTTCTCAATCTGCTTCTGAATTTCC
Phenotype
MIM: 300314
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAF7L PubMed Links

Gene Details

Gene
TAF7L
Gene Name
TATA-box binding protein associated factor 7 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168474.1 1059 Missense Mutation AAG,GAG K315E NP_001161946.1
NM_024885.3 1059 Missense Mutation AAG,GAG K401E NP_079161.3
XM_005262152.1 1059 Missense Mutation AAG,GAG K316E XP_005262209.1
XM_006724664.1 1059 Missense Mutation AAG,GAG K402E XP_006724727.1

View Full Product Details