Product Details

SNP ID
rs201485639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:12726653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCCAAGATTGGTTTCCACTTCTA[C/G]AACTTCTCGCCATGGCCTTAAATCC
Phenotype
MIM: 400005
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
USP9Y PubMed Links

Gene Details

Gene
USP9Y
Gene Name
ubiquitin specific peptidase 9, Y-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004654.3 1462 Missense Mutation CAA,GAA Q173E NP_004645.2
XM_017030078.1 1462 Missense Mutation CAA,GAA Q173E XP_016885567.1

View Full Product Details