Product Details
- SNP ID
-
rs202165661
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:119637085 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGCCCATCTCCTCCAGCTTACAGC[A/G]GCGATACAGCTCATAGTGCCGGGTG
- Phenotype
-
MIM: 300683
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR766
PubMed Links
Gene Details
- Gene
- MIR766
- Gene Name
- microRNA 766
There are no transcripts associated with this gene.
- Gene
- SEPT6
- Gene Name
- septin 6
View Full Product Details