Product Details

SNP ID
rs202093233
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:120626959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGTTTTACAAGGATAATACAGT[A/G]TACTCGAAAGCTCTTACTGAGCTCC
Phenotype
MIM: 300611 MIM: 300587
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1GALT1C1 PubMed Links

Gene Details

Gene
C1GALT1C1
Gene Name
C1GALT1 specific chaperone 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011551.2 447 Missense Mutation CAC,TAC H70Y NP_001011551.1
NM_152692.4 447 Missense Mutation CAC,TAC H70Y NP_689905.1
Gene
MCTS1
Gene Name
MCTS1, re-initiation and release factor
There are no transcripts associated with this gene.

View Full Product Details