Product Details

SNP ID
rs202070325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:119087235 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCTATCTGAGGCTTGGGAAGAA[C/T]GTTTGGGAGGCAGCTGCTTTTTAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA1210 PubMed Links

Gene Details

Gene
KIAA1210
Gene Name
KIAA1210
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020721.1 3584 Missense Mutation CAT,CGT H1332R NP_065772.1
XM_006724774.3 3584 Missense Mutation CAT,CGT H1156R XP_006724837.1
XM_017029688.1 3584 Missense Mutation CAT,CGT H1171R XP_016885177.1
XM_017029689.1 3584 Missense Mutation CAT,CGT H1105R XP_016885178.1

View Full Product Details