Product Details

SNP ID
rs200534434
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:83873208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAAAACTCCCTTAAAATCATCAC[A/G]TGAAAATGAACAATCCAAGAAGTCA
Phenotype
MIM: 300768
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYLC1 PubMed Links

Gene Details

Gene
CYLC1
Gene Name
cylicin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271680.1 554 Intron NP_001258609.1
NM_021118.2 554 Missense Mutation CAT,CGT H167R NP_066941.1
XM_005262086.3 554 Missense Mutation CAT,CGT H166R XP_005262143.1

View Full Product Details